missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran, Spectrophotometric Grade, >99.7%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32468K7

 View more versions of this product

Catalog No. AA32468K7



Specifications

Specifications

Tetrahydrofuran
109-99-9
Colorless liquid
C4H8O
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
Ether-like
Colorless
Unstable
Spectrophotometric grade
>99.7%
MFCD00005356
C1CCOC1
72.107
CHEBI:26911
Liquid
200 hPa (150mm Hg) at 20°C
4L

Chemical Identifiers

109-99-9
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
CHEBI:26911
C1CCOC1
C4H8O
MFCD00005356
8028
oxolane
Safety and Handling

Safety and Handling

H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-726-8

SDS Documents