Please login to your online account to display your discounted pricing

Tetrahydrofuran, Packaged under Argon, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$258.33 - $818.28

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Name Note Packaged under Argon
Purity Grade HPLC Grade
Molecular Formula C4H8O
Formula Weight 72.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004418

 
avantor performance
944103
Narrow Mouth Amber Glass Bottle 4L Each for $673.04

02004417

 
avantor performance
944102
Narrow Mouth Amber Glass Bottle 1L Each for $258.33

02004419

 
avantor performance
944133
Narrow Mouth PVC Coated Amber Glass Bottle 4L Each for $818.28
Description & Specifications

Specifications

Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.88kg
Name Note Packaged under Argon
Purity Grade HPLC Grade
Flash Point −14.00°C
Quantity 4L
Molecular Formula C4H8O
CAS 109-99-9

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry