Learn More
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
$169.75 - $547.03
Chemical Identifiers
CAS | 109-99-9 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005356 |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
IUPAC Name | oxolane |
SMILES | C1CCOC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA38994K2
|
Thermo Scientific Chemicals
038994K2 |
1 L |
Each for $169.75
|
|
|||||
AA38994M1
|
Thermo Scientific Chemicals
038994M1 |
4 x 1 L |
Each for $547.03
|
|
|||||
AA38994K7
|
Thermo Scientific Chemicals
038994K7 |
4 L |
Each for $397.08
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Specifications
°C to 108°C | |
Colorless | |
0.889 | |
1 L | |
Ether-like | |
1.407 | |
C4H8O | |
200 hPa (150mm Hg) at 20°C | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
Tetrahydrofuran, With 250ppm BHT |
109-99-9 | |
Colorless liquid | |
66°C | |
-17°C (1°F) | |
Liquid | |
>99.7% | |
MFCD00005356 | |
102391 | |
Miscible with water. | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
HPLC |
Safety and Handling
Keep away from food, drink and animal feeding stuffs. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H225-H302-H319-H335-H336-H351
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 203-726-8
RTECSNumber : LU5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only