Please login to your online account to display your discounted pricing

Tetrahydrofuran (Histological), Fisher Chemical


Contains about 0.025% butylated hydroxytoluene as a preservative

Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  T4004

 View more versions of this product

Catalog No. T400-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 72.11
Purity Grade Histological Grade
Boiling Point 66°C
Melting Point −108.4°C
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless
Vapor Pressure 200mbar at 20°C
Viscosity 0.55 cP at 20°C
pH 7 to 8
Density 0.880g/cm³
Water 1% max.
CAS 128-37-0
Cleanup Symbol Solvent
Molecular Formula C4H8O
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1
Linear Formula CH2(CH2)2CH2O
Chemical Name or Material Tetrahydrofuran
Identification Pass Test
Packaging Glass Bottle
Residue after Evaporation Pass Test
MDL Number MFCD00005356
CAS 109-99-9
Synonym THF
Dilution Test Pass Test

Specifications

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
CAS 128-37-0
Purity Grade Histological Grade
Molecular Formula C4H8O
Linear Formula CH2(CH2)2CH2O
Formula Weight 72.11
MDL Number MFCD00005356
Synonym THF
Accessories

Accessories for Catalog Number: T4004

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1

DANGER!

 

SDS & Safety Documents