Please login to your online account to display your discounted pricing

Tetrahydrofuran (Histological), Fisher Chemical


Contains about 0.025% butylated hydroxytoluene as a preservative

Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  T40020

 View more versions of this product

Catalog No. T400-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Histological Grade
Boiling Point 66°C
Melting Point −108.4°C
Formula Weight 72.11
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 200mbar at 20°C
Viscosity 0.55 cP at 20°C
Physical Form Liquid
Color Colorless
pH 7 to 8
Density 0.880g/cm³
Packaging Steel Pail
Water 1% max.
CAS 128-37-0
Synonym THF
Dilution Test Pass Test
Chemical Name or Material Tetrahydrofuran
Identification Pass Test
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1
Molecular Formula C4H8O
ChemAlert Storage Symbol Red
Cleanup Symbol Solvent
Residue after Evaporation Pass Test
Linear Formula CH2(CH2)2CH2O
MDL Number MFCD00005356
CAS 109-99-9

Specifications

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
CAS 128-37-0
Purity Grade Histological Grade
Molecular Formula C4H8O
Linear Formula CH2(CH2)2CH2O
Formula Weight 72.11
MDL Number MFCD00005356
Synonym THF
Accessories
 

Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1

DANGER!

 

SDS & Safety Documents