Please login to your online account to display your discounted pricing

Tetrahydrofuran For spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$188.89 - $709.00

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Molecular Formula C4H8O
Formula Weight 72.11
UN Number 2056
Synonym THF, Tetramethylene oxide, Oxolane, Butylene oxide, Oxolane, Butylene oxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1081100500

 
emd millipore corporation
1081100500
Glass Bottle 500mL Each for $188.89

M1081102500

 
emd millipore corporation
1081102500
Glass Bottle 2.5L Each for $709.00
Description & Specifications

Specifications

Boiling Point 65° to 66°C
Melting Point −108°C
Formula Weight 72.11
pH 7 to 8 (200g/L, H2O, at 20°C)
Quantity 500mL
Health Hazard 3 H335: May cause respiratory irritation.
Molecular Formula C4H8O
Color Colorless
CAS 109-99-9
UN Number 2056
Synonym THF, Tetramethylene oxide, Oxolane, Butylene oxide, Oxolane, Butylene oxide
Chemical Name or Material Tetrahydrofuran
Ignition Point 215°C (419°F)
Odor Ether-like
Packaging Glass Bottle
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protect from light.
Vapor Pressure 173 hPa (20°C)
Flash Point −20°C (−4°F)
Physical Form Liquid
Solubility Information Soluble in water at 20°C
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H319: Causes serious eye irritation.
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Density 0.89g/cm3 at 20°C
Safety and Handling

H225: Highly flammable liquid and vapour.

H335: May cause respiratory irritation.

H319: Causes serious eye irritation.