Please login to your online account to display your discounted pricing

Tetrahydrofuran-D8, deuteration degree min 99.5% for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$98.03 - $699.13

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran-D8
CAS 1693-74-9
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C4D8O (Hill Formula)
Formula Weight 80.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1133640001

 
emd millipore corporation
1133640001
Glass ampoule 1mL Each for $98.03

M1133640009

 
emd millipore corporation
1133640009
Glass ampoule 10 x 0.75mL Each for $586.48

M1133640010

 
emd millipore corporation
1133640010
Septum ampoule 10mL Each for $699.13
Description & Specifications

Specifications

Formula Weight 80.16
Quantity 1mL
Color Colorless
Physical Form Liquid
Packaging Glass ampoule
Odor Ether-like
CAS 1693-74-9
Molecular Formula C4D8O (Hill Formula)
Assay Percent Range 99.5% min.
Chemical Name or Material Tetrahydrofuran-D8