Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofuran-d8, 99.5% (Isotopic)

$77.59 - $77.59

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran-d8
CAS 1693-74-9
Name Note Isotopic
Formula Weight 80.16
MDL Number MFCD00044238
UN Number UN2056
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3655103

  SDS
alfa aesar™
3655103
1g Each for $77.59
Description & Specifications

Specifications

Formula Weight 80.16
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless
Health Hazard 2 GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 66°C
Flash Point −17° (−1°F)
CAS 1693-74-9
Quantity 1g
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
Storage Note 1 2°C to 8°C
MDL Number MFCD00044238
Chemical Name or Material Tetrahydrofuran-d8
Assay 99.5%
Density 0.985g/mL
Melting Point −106°C
Name Note Isotopic
Health Hazard 1 Danger
UN Number UN2056
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C