missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran ChromAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK28584 Avantor Performance
285806
Glass 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

109-99-9
C4H8O
72.11
Tetrahydrofuran

Specifications

109-99-9
72.11
4L
C4H8O
Glass
Tetrahydrofuran