Please login to your online account to display your discounted pricing

Tetrahydrofuran (Certified), Fisher Chemical


Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  T39720

 View more versions of this product

Catalog No. T397-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Boiling Point 66°C
Melting Point −108.4°C
Formula Weight 72.11
Quantity 20L
UN Number 2056
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 200mbar at 20°C
Viscosity 0.55 cP at 20°C
Physical Form Liquid
Color Colorless
pH 7 to 8
Density 0.880g/cm³
Packaging Steel Pail
Peroxides ≤5ppm
Water ≤0.02%
Molecular Formula C4H8O
MDL Number MFCD00005356
Boiling Range <2°C (1-95mL)
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1
Identification Pass Test
Specific Gravity 0.884 to 0.887 (at 25°C)
Specific Gravity at 25 degC Inclusive between 0.884°C
Stabilizer 75-400ppm
Purity Grade Notes Contains about 0.025% butylated hydroxytoluene as a preservative
CAS 128-37-0
Chemical Name or Material Tetrahydrofuran
Cleanup Symbol Solvent
CAS 109-99-9
Health Hazard 2 DANGER!

Specifications

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
CAS 128-37-0
Purity Grade Certified Grade
Purity Grade Notes Contains about 0.025% butylated hydroxytoluene as a preservative
Molecular Formula C4H8O
Formula Weight 72.11
MDL Number MFCD00005356
Accessories

Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1

DANGER!

 

SDS & Safety Documents