missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofuran, Biograde, 99.8%, unstab.

Nucleic acid and peptide synthesis

Manufacturer:  Alfa Aesar™ 44505K7

 View more versions of this product

Catalog No. AA44505K7



Specifications

Specifications

Tetrahydrofuran
109-99-9
MFCD00005356
102391
1,4-epoxybutane
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
99.8%
0.889 g/mL
1.407
66°C
-108°C
Unstabilized
C4H8O
UN2056
14,9211
Miscible with water.
C1CCOC1
72.107
CHEBI:26911
Liquid
Biograde
Ether-like
−17°C (1°F)
Colorless
4L

Chemical Identifiers

109-99-9
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
1,4-epoxybutane
CHEBI:26911
C1CCOC1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302-H319-H335-H336-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN

EINECSNumber : 203-726-8

RTECSNumber : LU5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents