Please login to your online account to display your discounted pricing

Tetrahydrofuran, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$246.57 - $1,122.67

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C4H8O
Formula Weight 72.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004421

 
avantor performance
945003
Narrow Mouth Amber Glass Bottle 4L Each for $924.94

02004422

 
avantor performance
945005
Aluminum Bottle 4L Each for $1,027.02

02004423

 
avantor performance
945033
Narrow Mouth PVC Coated Amber Bottle 4L Each for $1,122.67

02004424

 
avantor performance
945001
Narrow Mouth Amber Glass Bottle 500mL Each for $246.57
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Molecular Formula C4H8O
Quantity 4L
Flash Point −14.00°C
Specific Gravity 1L = 0.88kg
Packaging Narrow Mouth Amber Glass Bottle