Please login to your online account to display your discounted pricing

Tetrahydrofuran, anhydrous, Alfa Aesar™

$143.84 - $143.84

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Physical Form Clear colorless liquid
Assay Percent Range >99.8%
Molecular Formula C4H8O; M.P.: -108°C
Formula Weight 72.11g/mol
MDL Number MFCD00005356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45829K2

 
alfa aesar
45829-K2
1L Each for $143.84
Description & Specifications

Specifications

Density 0.889
Quantity 1L
Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Physical Form Clear colorless liquid
Assay Percent Range >99.8%
Molecular Formula C4H8O; M.P.: -108°C
Formula Weight 72.11g/mol
MDL Number MFCD00005356
EINECS Number 203-726-8