Learn More
Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal bottles, Thermo Scientific Chemicals
$83.84 - $202.92
Chemical Identifiers
CAS | 109-99-9 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005356 |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
IUPAC Name | oxolane |
SMILES | C1CCOC1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
CHEBI:26911 | |
C1CCOC1 |
C4H8O | |
MFCD00005356 | |
8028 | |
oxolane |
Specifications
0.889 g/mL | |
-17°C (1°F) | |
100 mL | |
Ether-like | |
109-99-9 | |
-108°C | |
MFCD00005356 | |
102391 | |
Miscible with water. | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
≥99.8% |
1.407 | |
Colorless | |
ChemSeal™ bottles | |
Liquid | |
66°C | |
C4H8O | |
UN2056 | |
14,9211 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
Tetrahydrofuran, BHT-free |
Safety and Handling
GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H225-H302-H319-H335-H336-H351
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
EINECSNumber : 203-726-8
RTECSNumber : LU5950000
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only