missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran, anhydrous, >99.8%%, Alfa Aesar™

For Synthesis and Liquid Chromatography

Manufacturer:  Thermo Fisher Scientific 44608M1

 View more versions of this product

Catalog No. AA44608M1



Specifications

Specifications

Tetrahydrofuran
109-99-9
>99.8%
MFCD00005356
1,4-epoxybutane
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
0.889
1.407
-17°C (1°F)
Colorless
4 x 1L
Water <100ppm
Colorless liquid
C4H8O
102391
Miscible with water.
C1CCOC1
72.107
CHEBI:26911
Liquid
Ether-like
200 hPa (150mm Hg) at 20°C
66°C
°C to 108°C

Chemical Identifiers

109-99-9
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
1,4-epoxybutane
CHEBI:26911
C1CCOC1
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302-H319-H335-H336-H351

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-726-8

RTECSNumber : LU5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents