missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran, ACS, >99%, Alfa Aesar™

For Synthesis and Liquid Chromatography

Manufacturer:  Alfa Aesar 30760M1

 View more versions of this product

Catalog No. AA30760M1



Description

Description

Maximum level of impurities: Color (APHA) 20, Peroxide (as H2O2) 0.015%, Residue after evaporation 0.03%, H2O 0.05%
Specifications

Specifications

Tetrahydrofuran
ACS
>99%
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
Ether-like
Colorless
109-99-9
Colorless liquid
C4H8O
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
C1CCOC1
72.107
CHEBI:26911
Liquid
200 hPa (150mm Hg) at 20°C
4 x 1L

Chemical Identifiers

109-99-9
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1
Safety and Handling

Safety and Handling

H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-726-8

SDS Documents