Please login to your online account to display your discounted pricing

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, ACROS Organics™


Quantity 5L
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  AC176630050

 View more versions of this product

Catalog No. AC176630050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Purity Grade Extra Pure
Formula Weight 72.11
Physical Form Liquid
Quantity 5L
Color Clear
Beilstein 17, 10
Merck Index 15, 9356
Packaging Plastic Bottle
Density 0.8800g/mL
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Residue after Evaporation 0.005% max.
Health Hazard 1 Danger
CAS 109-99-9
Molecular Formula C4H8O
Water 0.05% max. (Coulometric)
CAS 128-37-0
Peroxides 0.015% max. (as H2O2)
Stabilizer 75 to 400ppm BHT
Synonym THF
MDL Number MFCD00005356
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.
Chemical Name or Material Tetrahydrofuran
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.

Specifications

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
CAS 128-37-0
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C4H8O
Formula Weight 72.11
MDL Number MFCD00005356
Synonym THF
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.

SDS & Safety Documents