Please login to your online account to display your discounted pricing

Tetrahydrofuran, 99.9%, Extra Dry, Stabilized, AcroSeal™, ACROS Organics™

$136.80 - $136.80

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
Name Note Stabilized, water < 50ppm
CAS 109-99-9
CAS 128-37-0
Purity Grade Extra Dry
Assay ≥99.9%
Assay Percent Range 99.9%
Molecular Formula C4 H8 O
Formula Weight 72.11
MDL Number MFCD00005356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC610900010

  SDS
thermo fisher scientific
610900010
1L Each for $136.80
Description & Specifications

Specifications

Assay ≥99.9%
Formula Weight 72.11
Quantity 1L
EINECS Number 203-726-8
TSCA TSCA
Purity Grade Extra Dry
Density 0.8800g/mL
RTECS Number LU5950000
Beilstein 17, 10
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Chemical Name or Material Tetrahydrofuran
Additional Information Vapor Pressure: 200mbar at 20°C
Peroxides 5ppm max.
CAS 109-99-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Identification Pass Test
Stabilizer 75 - 400ppm
Name Note Stabilized, water < 50ppm
Water 50ppm max.
Assay Percent Range 99.9%
Molecular Formula C4 H8 O
MDL Number MFCD00005356
Synonym THF
CAS 128-37-0
Health Hazard 3 GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.