Please login to your online account to display your discounted pricing

Tetrahydrofuran, Spectrophotometric Grade 99.5+%, ACROS Organics™


Quantity 500mL

Manufacturer: thermo fisher scientific  AC222165000

 View more versions of this product

Catalog No. AC222165000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 72.11
Quantity 500mL
Color Clear
Assay ≥99.5%
Purity Grade For spectroscopy
Density 0.8800g/mL
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Peroxides ≤0.01%
Additional Information Vapor Pressure: 200mbar at 20°C
Alkalinity ≤0.0006meq/g
Absorbance (1 cm cell vs HPLC-grade water)
Synonym THF
Chemical Name or Material Tetrahydrofuran
Name Note For spectroscopy
CAS 109-99-9
Absorbance Conditions (1cm cell vs HPLC-grade water)
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses
Water ≤0.03%
Health Hazard 1 Danger
Identification Pass Test
Residue after Evaporation ≤0.005%
Molecular Formula C4H8O
CAS 128-37-0
Acidity ≤0.0002meq/g
Assay Percent Range 99.5+%
MDL Number MFCD00005356

Specifications

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
CAS 128-37-0
Purity Grade For spectroscopy
Assay ≥99.5%
Assay Percent Range 99.5+%
Molecular Formula C4H8O
Formula Weight 72.11
Synonym THF
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.

SDS & Safety Documents