Learn More
Tetrahydrofuran, 99.5%, for analysis, stabilized with BHT, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 446400025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrahydrofuran | |
109-99-9,128-37-0 | |
66.0°C | |
Glass bottle | |
99.4% min. (GC) | |
C4H8O | |
0.456 mPa.s (20°C) | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
Analytical |
-108.4°C | |
0.8800g/mL | |
2.5 L | |
1.407 | |
0.88 | |
MFCD00005356 | |
15,9356 | |
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
99.5% |
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspected of causing cancer.
May form
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 203-726-8
RUO – Research Use Only