Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofuran-3-one, 95%

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran-3-one
CAS 22929-52-8
Formula Weight 86.09
MDL Number MFCD07778393
Beilstein 105200
Synonym Dihydrofuran-3(2H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6694003

  SDS
alfa aesar™
H6694003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6694006

  SDS
alfa aesar™
H6694006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 86.09
CAS 22929-52-8
Chemical Name or Material Tetrahydrofuran-3-one
MDL Number MFCD07778393
Synonym Dihydrofuran-3(2H)-one
Beilstein 105200
Physical Form Liquid
Quantity 1g
Storage Note 1 2° to 8°C
Assay 95%
Safety and Handling

2° to 8°C