Please login to your online account to display your discounted pricing

Tetrahydro-β-carboline, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427580050

 View more versions of this product

Catalog No. AC427580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Crystals
Assay ≥97.5% (HPLC)
Formula Weight 172.23
Quantity 5g
Color White to slightly yellow
Packaging Glass bottle
Chemical Name or Material Tetrahydro-β-carboline
NMR Consistent with structure (1H-NMR)
Assay Percent Range 98%
Molecular Formula C11H12N2
MDL Number MFCD00004954
CAS 16502-01-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Infrared Spectrum Authentic
Synonym 1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Tetrahydro-β-carboline
CAS 16502-01-5
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C11H12N2
Formula Weight 172.23
MDL Number MFCD00004954
Synonym 1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole
Accessories

GHS Signal Word: Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Causes severe skin burns and eye damage.

SDS & Safety Documents