Please login to your online account to display your discounted pricing

Tetrahydro-4H-pyran-4-ol, Acros Organics

Chemical Identifiers

Chemical Name or Material Tetrahydro-4H-pyran-4-ol
CAS 2081-44-9
Molecular Formula C5H10O2
Formula Weight 102.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189430010

  SDS
thermo fisher scientific
AC189430010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC189430050

 
thermo fisher scientific
AC189430050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.13
Quantity 1g
Chemical Name or Material Tetrahydro-4H-pyran-4-ol
CAS 2081-44-9
Molecular Formula C5H10O2