missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Pyrazolidinone hydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 335490010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Pyrazolidinone hydrochloride | |
Authentic | |
Glass bottle | |
MFCD00043787 | |
3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride | |
HAYWTXQRYXYRAF-UHFFFAOYSA-N | |
pyrazolidin-3-one;hydrochloride | |
2733712 | |
98% |
1752-88-1 | |
98% | |
C3H7ClN2O | |
1g | |
Solubility in water: soluble | |
[H+].[Cl-].O=C1CCNN1 | |
122.55 | |
122.55 |
Chemical Identifiers
1752-88-1 | |
122.55 | |
HAYWTXQRYXYRAF-UHFFFAOYSA-N | |
2733712 | |
[H+].[Cl-].O=C1CCNN1 |
C3H7ClN2O | |
MFCD00043787 | |
3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride | |
pyrazolidin-3-one;hydrochloride |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only