Please login to your online account to display your discounted pricing

Tetrahydro-3-furoic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrahydro-3-furoic acid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H8O3
Formula Weight 116.12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC433450050

 
thermo fisher scientific
AC433450050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC433450250

 
thermo fisher scientific
AC433450250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 116.12
Quantity 5g
Color Colorless to pale yellow
Density 1.214g/mL
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Tetrahydro-3-furoic acid
Assay Percent Range 97%
Molecular Formula C5H8O3
CAS 89364-31-8
Infrared Spectrum Authentic
Stabilizer (250ppm BHT inhibitor)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.