Please login to your online account to display your discounted pricing

Tetrahydro-2H-thiopyran-4-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC61904DA

 View more versions of this product

Catalog No. CC61904DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 130.21
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Tetrahydro-2H-thiopyran-4-carbaldehyde
Packaging Amber Glass Bottle
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C6H10OS
MDL Number MFCD03425266
CAS 50675-19-9

Specifications

Chemical Name or Material Tetrahydro-2H-thiopyran-4-carbaldehyde
CAS 50675-19-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H10OS
Formula Weight 130.21
MDL Number MFCD03425266
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.