missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydro-2H-thiopyran-4-carbaldehyde, 97%, Maybridge

$325.00 - $325.00

Chemical Identifiers

CAS 50675-19-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H10OS
Formula Weight 130.21
MDL Number MFCD03425266
Chemical Name or Material Tetrahydro-2H-thiopyran-4-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC61904DA SDS Fisher Scientific Company
CC61904DA
Amber Glass Bottle 1g Each for $325.00
Specifications Description & Specifications

Chemical Identifiers

50675-19-9
100.0
C6H10OS
MFCD03425266
97%
96.5
97%
130.21
Tetrahydro-2H-thiopyran-4-carbaldehyde

Specifications

1g
96.5
Amber Glass Bottle
C6H10OS
MFCD03425266
97%
50675-19-9
100.0
97%
130.21
Tetrahydro-2H-thiopyran-4-carbaldehyde

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning