missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tetrahydro-2h-pyran-4-ylmethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC29909DA
Specifications
Tetrahydro-2H-pyran-4-ylmethanol | |
97% | |
C6H12O2 | |
1g | |
YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
(oxan-4-yl)methanol | |
2773573 | |
97% |
14774-37-9 | |
Amber Glass Bottle | |
MFCD00457804 | |
tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
OCC1CCOCC1 | |
116.16 | |
116.16 |
Chemical Identifiers
14774-37-9 | |
116.16 | |
YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
2773573 | |
OCC1CCOCC1 |
C6H12O2 | |
MFCD00457804 | |
tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
(oxan-4-yl)methanol |