Please login to your online account to display your discounted pricing

Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  CC29901EA

 View more versions of this product

Catalog No. CC29901EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 130.14
Quantity 10g
Assay 97%
CAS 1255400
Chemical Name or Material Tetrahydro-2H-pyran-4-carboxylic acid
Molecular Formula C6H10O3
Assay Percent Range 97%
Health Hazard 2 GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Specifications

Chemical Name or Material Tetrahydro-2H-pyran-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H10O3
Formula Weight 130.14
Accessories

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.