missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrahydro-2-furoic acid, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 263145000
Specifications
Tetrahydro-2-furoic acid | |
139°C | |
Glass bottle | |
1.17 | |
Yellow to Brown | |
99+% | |
99.0 | |
98% min. (GC) | |
MFCD00022473 | |
Solubility in water: soluble | |
C1CC(OC1)C(=O)O | |
116.12 | |
116.12 | |
98+% |
1.1700g/mL | |
Authentic | |
1.4590 to 1.4610 | |
128.0°C to 129.0°C (13.0mmHg) | |
500g | |
16874-33-2 | |
100.0 | |
C5H8O3 | |
2-furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, 2-tetrahydrofuroic acid, furancarboxylic acid, tetrahydro, pubchem7088, tetrahydro-2-furancarboxylic acid, tetrahydro-2-furoic acid, tetrahydrofuran-2-carboxylic acid, tetrahydrofuroic acid, thfc | |
UJJLJRQIPMGXEZ-UHFFFAOYSA-N | |
oxolane-2-carboxylic acid | |
86079 | |
Liquid |
Chemical Identifiers
16874-33-2 | |
116.12 | |
UJJLJRQIPMGXEZ-UHFFFAOYSA-N | |
86079 | |
C1CC(OC1)C(=O)O |
C5H8O3 | |
MFCD00022473 | |
2-furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, 2-tetrahydrofuroic acid, furancarboxylic acid, tetrahydro, pubchem7088, tetrahydro-2-furancarboxylic acid, tetrahydro-2-furoic acid, tetrahydrofuran-2-carboxylic acid, tetrahydrofuroic acid, thfc | |
oxolane-2-carboxylic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
TSCA : TSCA