missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2777403
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Tetrahydro-2,2-dimethyl-4H-pyran-4-one | |
60°C to 64°C (8 mmHg) | |
MFCD01549337 | |
2,2-dimethyltetrahydropyran-4-one, 2,2-dimethyldihydro-2h-pyran-4 3h-one, 2,2-dimethyl-tetrahydro-pyran-4-one, tetrahydro-2,2-dimethyl-4h-pyran-4-one, 2,2-dimethyltetrahydro-4h-pyran-4-one, 4h-pyran-4-one, tetrahydro-2,2-dimethyl, 2,2-dimethyl-2h-3,5,6-trihydropyran-4-one, pubchem24423, acmc-1bt7l | |
BWMNOXJVRHGUQM-UHFFFAOYSA-N | |
2,2-dimethyloxan-4-one | |
1738159 | |
95% |
1194-16-7 | |
C7H12O2 | |
1 g | |
Slightly soluble in water. | |
CC1(C)CC(=O)CCO1 | |
128.17 | |
128.17 |
Chemical Identifiers
1194-16-7 | |
128.17 | |
BWMNOXJVRHGUQM-UHFFFAOYSA-N | |
1738159 | |
CC1(C)CC(=O)CCO1 |
C7H12O2 | |
MFCD01549337 | |
2,2-dimethyltetrahydropyran-4-one, 2,2-dimethyldihydro-2h-pyran-4 3h-one, 2,2-dimethyl-tetrahydro-pyran-4-one, tetrahydro-2,2-dimethyl-4h-pyran-4-one, 2,2-dimethyltetrahydro-4h-pyran-4-one, 4h-pyran-4-one, tetrahydro-2,2-dimethyl, 2,2-dimethyl-2h-3,5,6-trihydropyran-4-one, pubchem24423, acmc-1bt7l | |
2,2-dimethyloxan-4-one |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only