Learn More
Tetrahexylammonium Bromide, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 420401000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalystSpecifications
Tetrahexylammonium bromide | |
Authentic | |
Glass bottle | |
[CH3(CH2)5]4NBr | |
100g | |
SYZCZDCAEVUSPM-UHFFFAOYSA-M | |
tetrahexylazanium;bromide | |
78026 | |
99% |
4328-13-6 | |
98.5% min. (Argentometry) | |
C24H52BrN | |
MFCD00011858 | |
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1 | |
[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC | |
434.59 | |
434.58 |
Chemical Identifiers
4328-13-6 | |
434.59 | |
SYZCZDCAEVUSPM-UHFFFAOYSA-M | |
78026 | |
[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC |
C24H52BrN | |
MFCD00011858 | |
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1 | |
tetrahexylazanium;bromide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 224-363-1
RUO â Research Use Only