Please login to your online account to display your discounted pricing

Tetraheptylammonium Iodide 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  420381000

 View more versions of this product

Catalog No. AC420381000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 537.7
Quantity 100g
Color White
TSCA TSCA
EINECS Number 222-570-1
Melting Point 122° to 126°C
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula (CH3(CH2)6)4NI
Molecular Formula C28 H60 I N
Chemical Name or Material Tetraheptylammonium iodide
MDL Number MFCD00041984
Name Note 99%
Assay Percent Range 99%
CAS 3535-83-9
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Tetraheptylammonium iodide
Name Note 99%
CAS 3535-83-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C28 H60 I N
Linear Formula (CH3(CH2)6)4NI
Formula Weight 537.7
MDL Number MFCD00041984
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents