Please login to your online account to display your discounted pricing

Tetraheptylammonium chloride, 95%, ACROS Organics™

$290.62 - $290.62

Chemical Identifiers

Chemical Name or Material Tetraheptylammonium chloride
Name Note 95%
CAS 10247-90-2
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C28H60ClN
Formula Weight 446.24
MDL Number MFCD00041983
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334540100

  SDS
thermo fisher scientific
334540100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC334540250

  SDS
thermo fisher scientific
334540250
Glass bottle 25g Each for $290.62
Description & Specifications

Specifications

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 446.24
Quantity 10g
Color Light yellow
EINECS Number 233-583-7
TSCA TSCA
Chemical Name or Material Tetraheptylammonium chloride
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%
Assay Percent Range 95%
Molecular Formula C28H60ClN
MDL Number MFCD00041983
CAS 10247-90-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Phase transfer catalyst

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.