Please login to your online account to display your discounted pricing

Tetraheptylammonium bromide, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC218161000

 View more versions of this product

Catalog No. AC218161000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Flakes
Formula Weight 490.69
Quantity 100g
Color White
Melting Point 89° to 91°C
Beilstein 04, IV, 736
Infrared Spectrum Authentic
Chemical Name or Material Tetraheptylammonium bromide
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C28H60BrN
Linear Formula [CH3(CH2)6]4NBr
MDL Number MFCD00011861
CAS 4368-51-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetraheptylammonium bromide
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C28H60BrN
Linear Formula [CH3(CH2)6]4NBr
Formula Weight 490.69
MDL Number MFCD00011861
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents