missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraformylhydrazine, 98%, ACROS Organics™

Chemical Identifiers

CAS 52023-52-6
Molecular Formula C4H4N2O4
Molecular Weight (g/mol) 144.086
MDL Number MFCD08059355
InChI Key NODMXZGWWULGLE-UHFFFAOYSA-N
Synonym 1,1,2,2-hydrazinetetracarbox aldehyde, hydrazinetetracarboxaldehyde, n,n',n'-triformylformohydrazide, tetraformylhydrazine
PubChem CID 11217319
IUPAC Name N-(diformylamino)-N-formylformamide
SMILES C(=O)N(C=O)N(C=O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC380480010
SDS View Documents
Thermo Fisher Scientific
AC380480010
1g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380480050
SDS View Documents
Thermo Fisher Scientific
AC380480050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

52023-52-6
144.086
NODMXZGWWULGLE-UHFFFAOYSA-N
11217319
C(=O)N(C=O)N(C=O)C=O
C4H4N2O4
MFCD08059355
1,1,2,2-hydrazinetetracarbox aldehyde, hydrazinetetracarboxaldehyde, n,n',n'-triformylformohydrazide, tetraformylhydrazine
N-(diformylamino)-N-formylformamide

Specifications

97.5% min.
Glass Bottle
180.0°C to 183.0°C
52023-52-6
C4H4N2O4
MFCD08059355
NODMXZGWWULGLE-UHFFFAOYSA-N
N-(diformylamino)-N-formylformamide
11217319
Powder
Tetraformylhydrazine
Authentic
White
1g
98%
[N(COH)2]2
1,1,2,2-hydrazinetetracarbox aldehyde, hydrazinetetracarboxaldehyde, n,n',n'-triformylformohydrazide, tetraformylhydrazine
C(=O)N(C=O)N(C=O)C=O
144.086
144.09
98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS