Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrafluoroterephthalonitrile, 98%

Chemical Identifiers

Chemical Name or Material Tetrafluoroterephthalonitrile
CAS 1835-49-0
Formula Weight 200.09
MDL Number MFCD00001776
UN Number UN3439
Synonym 2,3,5,6-Tetrafluoro-1,4-dicyanobenzene; 2,4,5,6-Tetrafluorobenzene-1,4-dicarbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6132614

  SDS
alfa aesar™
H6132614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 197° to 199°C
Formula Weight 200.09
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
RTECS Number CZ1955000
Health Hazard 1 Danger
Synonym 2,3,5,6-Tetrafluoro-1,4-dicyanobenzene; 2,4,5,6-Tetrafluorobenzene-1,4-dicarbonitrile
UN Number UN3439
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1835-49-0
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Physical Form Solid
MDL Number MFCD00001776
Quantity 25g
Chemical Name or Material Tetrafluoroterephthalonitrile

Applications
Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

Solubility
Soluble in hot methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.