Please login to your online account to display your discounted pricing

Tetrafluorophthalic anhydride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrafluorophthalic anhydride
CAS 652-12-0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8F4O3
Formula Weight 220.08g/mol
MDL Number MFCD00039697
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC367300050

  SDS
fisher scientific acros
367300050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Light gray to beige
Quantity 5g
Chemical Name or Material Tetrafluorophthalic anhydride
CAS 652-12-0
Physical Form Light grayish to beige powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8F4O3
Formula Weight 220.08g/mol
MDL Number MFCD00039697
Beilstein 17, V,11, 260
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.