Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrafluorophthalic acid, 97%

$118.84 - $118.84

Chemical Identifiers

Chemical Name or Material Tetrafluorophthalic acid
CAS 652-03-9
Molecular Formula C8H2F4O4
Formula Weight 238.1
MDL Number MFCD00002407
Beilstein 2057012
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2121406

 
alfa aesar™
B2121406
5g Each for $118.84
Description & Specifications

Specifications

Formula Weight 238.1
Chemical Name or Material Tetrafluorophthalic acid
Assay 97%
Molecular Formula C8H2F4O4
MDL Number MFCD00002407
CAS 652-03-9
Beilstein 2057012
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈156°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.