missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrafluoroisophthalonitrile, TCI America™

Manufacturer:  TCI America T172825G

 View more versions of this product

Catalog No. T172825G



Specifications

Specifications

Tetrafluoroisophthalonitrile
76°C
2377-81-3
MFCD00013290
WVHMPQKZPHOCRD-UHFFFAOYSA-N
2,4,5,6-tetrafluorobenzene-1,3-dicarbonitrile
608182
Crystal-Powder at 20°C
White-Yellow
25g
C8F4N2
tetrafluoroisophthalonitrile, 2,4,5,6-tetrafluoroisophthalonitrile, perfluoroisophthalonitrile, tetrafluorobenzene-1,3-dicarbonitrile, 1,3-benzenedicarbonitrile, 2,4,5,6-tetrafluoro, tetrafluoroisopthalonitrile, m-dicyanotetrafluorobenzene, acmc-209g5p, 1,3-dicyanoperfluorobenzene, tetrafluoroisophthalonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)C#N)F
200.096
200.10
≥98.0% (GC)

Chemical Identifiers

2377-81-3
200.096
WVHMPQKZPHOCRD-UHFFFAOYSA-N
608182
C(#N)C1=C(C(=C(C(=C1F)F)F)C#N)F
C8F4N2
MFCD00013290
tetrafluoroisophthalonitrile, 2,4,5,6-tetrafluoroisophthalonitrile, perfluoroisophthalonitrile, tetrafluorobenzene-1,3-dicarbonitrile, 1,3-benzenedicarbonitrile, 2,4,5,6-tetrafluoro, tetrafluoroisopthalonitrile, m-dicyanotetrafluorobenzene, acmc-209g5p, 1,3-dicyanoperfluorobenzene, tetrafluoroisophthalonitrile
2,4,5,6-tetrafluorobenzene-1,3-dicarbonitrile
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3882

TSCA : No

Documents