missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrafluoroboric acid-diethyl ether complex, 50-55% w/w HBF4

Manufacturer:  Alfa Aesar™ 18208AK

 View more versions of this product

Catalog No. AA18208AK



Specifications

Specifications

Tetrafluoroboric acid-diethyl ether complex
36° (97°F)
250mL
C4H11BF4O
MFCD00011345
14,4149
50 to 55% w/w HBF4
XFHGDBFMJCLEOW-UHFFFAOYSA-N
ethoxyethane;trifluoroborane;hydrofluoride
11344169
Liquid
1.19g/mL
Colorless to Orange
67969-82-8
HBF4·(CH3CH2)2O
UN2920
fluoroboric acid diethyl ether complex, tetrafluoroboric acid diethyl ether complex, hbf4.oet2, fluoroboric acid diethylether complex, boron trifluoride diethyl ether hydrofluoride
Reacts in water
B(F)(F)F.CCOCC.F
161.935
161.93

Chemical Identifiers

67969-82-8
161.935
XFHGDBFMJCLEOW-UHFFFAOYSA-N
11344169
B(F)(F)F.CCOCC.F
C4H11BF4O
MFCD00011345
fluoroboric acid diethyl ether complex, tetrafluoroboric acid diethyl ether complex, hbf4.oet2, fluoroboric acid diethylether complex, boron trifluoride diethyl ether hydrofluoride
ethoxyethane;trifluoroborane;hydrofluoride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H336
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : TSCA

SDS Documents