missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethyltin, 98%

Manufacturer:  Alfa Aesar™ 7114418

 View more versions of this product

Catalog No. AA7114418



Specifications

Specifications

Tetraethyltin
-112°C
50g
Characteristic
1.4730
MFCD00009020
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, tetraethyltin, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
RWWNQEOPUOCKGR-UHFFFAOYSA-N
tetraethylstannane
11704
Liquid
181°C
597-64-8
53°C (128°F)
1.187g/mL
C8H20Sn
UN3384
Insoluble in water; Soluble in alcohol and ether
CC[Sn](CC)(CC)CC
234.958
234.94
98%

Chemical Identifiers

597-64-8
234.958
RWWNQEOPUOCKGR-UHFFFAOYSA-N
11704
CC[Sn](CC)(CC)CC
C8H20Sn
MFCD00009020
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, tetraethyltin, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
tetraethylstannane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.

GHS P Statement
P273-P303+P361+P353-P309-P310-P501a
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

RTECSNumber : WH8625000

TSCA : TSCA

SDS Documents