Please login to your online account to display your discounted pricing

Tetraethyltin, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetraethyltin
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H20Sn
Linear Formula (C2H5)4Sn
Formula Weight 234.94
MDL Number MFCD00009020
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC212070050

  SDS
thermo fisher scientific
AC212070050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.94
Physical Form Liquid
Assay ≥96.0%
Quantity 5mL
Color Clear colorless
Refractive Index 1.4720 to 1.4740 (20°C, 589nm)
Assay Percent Range 97%
Linear Formula (C2H5)4Sn
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Tetraethyltin
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
CAS 597-64-8
Molecular Formula C8H20Sn
MDL Number MFCD00009020
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.