Please login to your online account to display your discounted pricing

Tetraethyltin, 97%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC212070050

 View more versions of this product

Catalog No. AC212070050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 234.94
Quantity 5mL
Color Clear colorless
Refractive Index 1.4720 to 1.4740 (20°C, 589nm)
Assay Percent Range 97%
Chemical Name or Material Tetraethyltin
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H20Sn
Linear Formula (C2H5)4Sn
MDL Number MFCD00009020
CAS 597-64-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetraethyltin
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H20Sn
Linear Formula (C2H5)4Sn
Formula Weight 234.94
MDL Number MFCD00009020
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.

SDS & Safety Documents