Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylsilane, 97%

Chemical Identifiers

Chemical Name or Material Tetraethylsilane
CAS 631-36-7
Molecular Formula C8H20Si
Formula Weight 144.33
MDL Number MFCD00009019
UN Number UN1993
Beilstein 1732768
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0332806

 
alfa aesar™
L0332806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0332814

 
alfa aesar™
L0332814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0332822

 
alfa aesar™
L0332822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.33
Chemical Name or Material Tetraethylsilane
Odor Recognizable
Assay 97%
Molecular Formula C8H20Si
MDL Number MFCD00009019
CAS 631-36-7
UN Number UN1993
Beilstein 1732768
Melting Point −82°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number VV5500000
TSCA TSCA
Boiling Point 153° to 155°C
Quantity 5g
Flash Point 26°C (78°F)

Applications
Used as the intermediates of OLED, pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS H Statement
H226
Flammable liquid and vapor.