missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Tetraethylpropylene-1,3-diphosphonate, 97%, ACROS Organics™
Chemical Identifiers
CAS |
22401-25-8 |
Molecular Formula |
C11H26O6P2 |
Molecular Weight (g/mol) |
316.27 |
MDL Number |
MFCD00048543 |
InChI Key |
LJQJJGQRJQSYQN-UHFFFAOYSA-N |
Synonym |
1,3-bis diethoxyphosphinyl propane, 1,3-bis diethoxyphosphoryl propane, phosphonic acid, 1,3-propanediylbis-, tetraethyl ester, phosphonic acid, trimethylenedi-, tetraethyl ester, propane-1,3-diphosphonic acid, tetraethyl ester, propanediyl-bis-phosphonic acid tetraethyl ester, tetraethyl 1,3-propylene bisphosphonate, tetraethyl propane-1,3-diphosphonate, tetraethyl propylenediphosphonate, tetraethyltrimethylenediphosphonate |
PubChem CID |
519945 |
IUPAC Name |
1,3-bis(diethoxyphosphoryl)propane |
SMILES |
CCOP(=O)(CCCP(=O)(OCC)OCC)OCC |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
AC349060050
SDS
View Documents
|
Thermo Fisher Scientific
349060050
|
5g |
Glass bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
Description
Description
For C-C Bond Formation (Olefination)
Specifications
Chemical Identifiers
|
22401-25-8 |
|
316.27 |
|
LJQJJGQRJQSYQN-UHFFFAOYSA-N |
|
519945 |
|
CCOP(=O)(CCCP(=O)(OCC)OCC)OCC |
|
C11H26O6P2 |
|
MFCD00048543 |
|
1,3-bis diethoxyphosphinyl propane, 1,3-bis diethoxyphosphoryl propane, phosphonic acid, 1,3-propanediylbis-, tetraethyl ester, phosphonic acid, trimethylenedi-, tetraethyl ester, propane-1,3-diphosphonic acid, tetraethyl ester, propanediyl-bis-phosphonic acid tetraethyl ester, tetraethyl 1,3-propylene bisphosphonate, tetraethyl propane-1,3-diphosphonate, tetraethyl propylenediphosphonate, tetraethyltrimethylenediphosphonate |
|
1,3-bis(diethoxyphosphoryl)propane |
Specifications
|
Authentic |
|
1.4450 to 1.4470 |
|
Colorless |
|
22401-25-8 |
|
C11H26O6P2 |
|
1,3-bis diethoxyphosphinyl propane, 1,3-bis diethoxyphosphoryl propane, phosphonic acid, 1,3-propanediylbis-, tetraethyl ester, phosphonic acid, trimethylenedi-, tetraethyl ester, propane-1,3-diphosphonic acid, tetraethyl ester, propanediyl-bis-phosphonic acid tetraethyl ester, tetraethyl 1,3-propylene bisphosphonate, tetraethyl propane-1,3-diphosphonate, tetraethyl propylenediphosphonate, tetraethyltrimethylenediphosphonate |
|
CCOP(=O)(CCCP(=O)(OCC)OCC)OCC |
|
316.27 |
|
316.27 |
|
97% |
|
Glass bottle |
|
206.0°C to 227.0°C (5.0mmHg) |
|
5g |
|
96% min. (GC) |
|
MFCD00048543 |
|
LJQJJGQRJQSYQN-UHFFFAOYSA-N |
|
1,3-bis(diethoxyphosphoryl)propane |
|
519945 |
|
Liquid |
|
Tetraethylpropylene-1, 3-diphosphonate, 97% |
SDS