missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylphosphonium Tetrafluoroborate, TCI America™

Manufacturer:  TCI America T174725G

 View more versions of this product

Catalog No. T174725G



Specifications

Specifications

Tetraethylphosphonium Tetrafluoroborate
25g
C8H20BF4P
1759
[B-](F)(F)(F)F.CC[P+](CC)(CC)CC
234.025
234.03
≥97.0% (T)
White
665-49-6
MFCD01631314
IOQILLMTUOVXHI-UHFFFAOYSA-N
tetraethylphosphanium;tetrafluoroborate
16218631
Crystal-Powder at 20°C

Chemical Identifiers

665-49-6
234.025
IOQILLMTUOVXHI-UHFFFAOYSA-N
tetraethylphosphanium;tetrafluoroborate
C8H20BF4P
MFCD01631314
16218631
[B-](F)(F)(F)F.CC[P+](CC)(CC)CC
Safety and Handling

Safety and Handling

TSCA : No

Documents