missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetraethyleneglycol monomethyl ether, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 420350250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tetraethyleneglycol monomethyl ether | |
23783-42-8 | |
158°C to 160°C (5.0mmHg) | |
Authentic | |
Glass bottle | |
1.4440 to 1.446 | |
MFCD00041756 | |
0.987 | |
ZNYRFEPBTVGZDN-UHFFFAOYSA-N | |
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanol | |
90263 | |
98% |
98% | |
0.9870g/mL | |
>110°C | |
98% | |
C9H20O5 | |
CH3O(CH2CH2O)3CH2CH2OH | |
25g | |
2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol, tetraethyleneglycol monomethylether, mpeg4-oh, 2-2-2-2-methoxy-ethoxy-ethoxy-ethoxy-ethanol, 2-2-2-2-methoxyethoxy ethoxy ethoxy ethanol, m-peg4-alcohol | |
COCCOCCOCCOCCO | |
208.25 | |
208.25 |
Safety and Handling
EINECSNumber : 245-883-5
TSCA : TSCA
RUO â Research Use Only