missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylene Glycol Dimethyl Ether, TCI America™

$24.44 - $68.75

Chemical Identifiers

CAS 143-24-8
Molecular Formula C10H22O5
Molecular Weight (g/mol) 222.281
MDL Number MFCD00008505
InChI Key ZUHZGEOKBKGPSW-UHFFFAOYSA-N
Synonym tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
PubChem CID 8925
ChEBI CHEBI:46785
IUPAC Name 1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
SMILES COCCOCCOCCOCCOC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B049725ML View Documents TCI America
B049725ML
25mL Each for $24.44
 
B0497500ML View Documents TCI America
B0497500ML
500mL Each for $68.75
 
Specifications

Chemical Identifiers

143-24-8
222.281
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
8925
1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
C10H22O5
MFCD00008505
tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
CHEBI:46785
COCCOCCOCCOCCOC

Specifications

276°C
25mL
C10H22O5
tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
COCCOCCOCCOCCOC
222.281
CHEBI:46785
Clear Liquid at 20°C
Tetraethylene Glycol Dimethyl Ether
Colorless
143-24-8
MFCD00008505
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
8925
222.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (7)-1321

RTECSNumber : SB0400000

TSCA : Yes