missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylene glycol dimethyl ether, 98+%

Manufacturer:  Alfa Aesar™ 2044330

 View more versions of this product

Catalog No. AA2044330



Specifications

Specifications

Tetraethylene glycol dimethyl ether
140° (284°F)
275°C to 276°C
250g
C10H22O5
14,9208
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
8925
222.28
≥98%
1.01g/mL
1.4330
-30°C
143-24-8
MFCD00008505
tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
COCCOCCOCCOCCOC
222.281
CHEBI:46785
Liquid

Chemical Identifiers

143-24-8
222.281
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
8925
1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
C10H22O5
MFCD00008505
tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
CHEBI:46785
COCCOCCOCCOCCOC
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : SB0400000

TSCA : TSCA

SDS Documents